Learn More
ERCC1 Mouse anti-Human, Rat, Clone: 2E12, lyophilized, UltraMAB™

Description
Specifications
Specifications
| Antigen | ERCC1 |
| Applications | Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Peptide Array |
| Classification | Monoclonal |
| Clone | 2E12 |
| Conjugate | Unconjugated |
| Formulation | PBS with 8% trehalose and no preservative |
| Gene | ERCC1 |
| Gene Accession No. | P07992 |
| Gene Alias | COFS4, RAD10, UV20 |
| Gene Symbols | Ercc1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.