Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ eRF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA586994
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA586994 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA586994 Supplier Invitrogen™ Supplier No. PA586994
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Purity is > 95% by SDS-PAGE.

Termination of protein biosynthesis and release of the nascent polypeptide chain are signaled by the presence of an in-frame stop codon at the aminoacyl site of the ribosome. The process of translation termination is universal and is mediated by protein release factors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen eRF1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene ETF1
Gene Accession No. P62495, Q5U2Q7, Q8BWY3
Gene Alias AI463371; D5S1995; D6Ertd109e; ERF; ERF1; ETF1; Eukaryotic peptide chain release factor subunit 1; eukaryotic release factor 1; eukaryotic translation termination factor 1; polypeptide chain release factor 1; protein Cl1; RF1; sup45 (yeast omnipotent suppressor 45) homolog-like 1; SUP45L1; TB3-1
Gene Symbols ETF1
Host Species Rabbit
Immunogen Recombinant fusion protein of human ETF1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2107, 225363, 307503
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.