Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ eRF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596476
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596476 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596476 Supplier Invitrogen™ Supplier No. PA596476
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VKFIQEKKLI GRYFDEISQD TGKYCFGVED TLKALEMGAV EILIVYENLD IMRYVLHCQG TEEEKILYLT PEQEKDKSHF TDKETGQEHE LIESMPLLEW FANNYKKFGA TLEIVTDKSQ EGSQFVKGFG GIGGILRYRV DFQGMEYQGG DDEFFDLDDY; Positive Samples: Raji, U-87MG, HT-1080, SKOV3, Mouse liver, Mouse heart, Mouse kidney, Rat heart; Cellular Location: Cytoplasm.

Termination of protein biosynthesis and release of the nascent polypeptide chain are signaled by the presence of an in-frame stop codon at the aminoacyl site of the ribosome. The process of translation termination is universal and is mediated by protein release factors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen eRF1
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.24 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ETF1
Gene Accession No. P62495, Q5U2Q7, Q8BWY3
Gene Alias AI463371; D5S1995; D6Ertd109e; ERF; ERF1; ETF1; Eukaryotic peptide chain release factor subunit 1; eukaryotic release factor 1; eukaryotic translation termination factor 1; polypeptide chain release factor 1; protein Cl1; RF1; sup45 (yeast omnipotent suppressor 45) homolog-like 1; SUP45L1; TB3-1
Gene Symbols ETF1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 278-437 of human ETF1 (NP_0047211).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2107, 225363, 307503
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.