Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ERGIC1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. p-7206904 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-0719 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-0719 Supplier Proteintech Group Inc Supplier No. 161081AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The MW of this protein is 32 kDa, and this antibody specifically recognizes the 32 kDa protein.

The function of ERGIC1 remains largely unknown. ERGIC1 was highly expressed in most primary prostate tumors. It is an intriguing potential drug target especially for the ERG oncogene expressing tumors.

Specifications

Antigen ERGIC1
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ERGIC1
Gene Accession No. Q969X5, Q9DC16
Gene Alias ERGIC 32, ERGIC1, ERGIC32, KIAA1181, NET24
Gene Symbols Ergic1
Host Species Rabbit
Immunogen ERGIC1 Fusion Protein Ag9025
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 287177, 57222, 67458
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.