Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ERGIC3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109263
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109263 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109263 Supplier Invitrogen™ Supplier No. PA5109263
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: Siha, human placenta tissue, rat bone marrow tissue, LOVO, human colon cancer tissue, human uterus tissue, mouse brain tissue.

ERGIC3 is possible role in transport between endoplasmic reticulum and Golgi.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ERGIC3
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene ERGIC3
Gene Accession No. Q9Y282
Gene Alias 2310015B14Rik; AV318804; C20orf47; CGI-54; D2Ucla1; dJ477O4.2; Endoplasmic reticulum-Golgi intermediate compartment protein 3; endoplasmic reticulum-localized protein ERp43; ERGIC and golgi 3; ERGIC3; ERV46; NY-BR-84; PRO0989; Sdbcag84; serologically defined breast cancer antigen 84; serologically defined breast cancer antigen NY-BR-84; Serologically defined breast cancer antigen NY-BR-84 homolog
Gene Symbols ERGIC3
Host Species Rabbit
Immunogen Synthetic peptide within Human ERGI3 aa 334-383.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51614
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles, store in dark
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.