Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NoTrax™ Ergo Trax™

Catalog No. 19085916 Shop All NoTrax Products
Change view
Click to view available options
Dimensions (L x W):
2 x 3 ft.
3 x 5 ft.
3 x 12 ft.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
19-085-916 3 x 5 ft.
19-085-915 2 x 3 ft.
19-085-917 3 x 12 ft.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-085-916 Supplier NoTrax™ Supplier No. 485S0035YB
Add to Cart
Add to Cart

Ergo Trax™ is engineered for industrial applications and is particularly suited for multi-shift operations in dry work areas

  • For industrial applications, multi-shift operations in dry work areas
  • Durable vinyl diamond-plate top surface design provides non-directional traction
  • Easy to clean
  • Top surface combined with Niru™ rubber sponge base utilizing NoTrax™ exclusive UniFusion™ technology
    • Eliminates possibility of de-lamination
  • Sloped beveled edges reduce tripping hazards and allow easy access on and off mat

Specifications

Type Anti-fatigue Floor Mat
Dimensions (L x W) 3 x 5 ft.
Color Black, Yellow
Length (English) 3 ft.
Material Heavy duty vinyl top surface combined with Niru™ rubber base
Width (English) 5 ft.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.