Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ ProFlex™ 600/615/620 Knee Sleeves

Catalog No. 19037835A Shop All Ergodyne Products
Change view
Click to view available options
Design:
Contoured
Contoured; Open patella/anterior pad
Neoprene four-way stretch Fabric, closed cell structure
Single layer, Neoprene four-way stretch Fabric
Size:
Small
Medium
Large
X-Large
2X-Large
Model:
600
615
620
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Design Size Model
19-037835A Single layer, Neoprene four-way stretch Fabric Small 600
19-037-835B Single layer, Neoprene four-way stretch Fabric Medium 600
19-037-835C Single layer, Neoprene four-way stretch Fabric Large 600
19-037-835D Contoured X-Large 600
19-037-835E Single layer, Neoprene four-way stretch Fabric 2X-Large 600
19-037-838D Contoured; Open patella/anterior pad X-Large 615
19-037-838B Contoured; Open patella/anterior pad Medium 615
19-037-838E Contoured; Open patella/anterior pad 2X-Large 615
19-037-838A Contoured; Open patella/anterior pad Small 615
19-037-839E Neoprene four-way stretch Fabric, closed cell structure 2X-Large 620
19-037-838C Contoured; Open patella/anterior pad Large 615
19-037-8398 Neoprene four-way stretch Fabric, closed cell structure Medium 620
19-037-839C Neoprene four-way stretch Fabric, closed cell structure Large 620
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Catalog No. 19-037835A Supplier Ergodyne™ Supplier No. 16502
Add to Cart
Add to Cart

Specifications

Color Black
Product Type Proflex™ Knee Sleeve
Design Single layer, Neoprene four-way stretch Fabric
Size Small
For Use With (Application) Provide glove-like fit and comfortable wear and provides uniform support and warmth to affected area
Material Neoprene
Model 600
Size (English) 13 to 14 in.
Size (Metric) 33 to 36 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.