Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ ProFlex™ 4000/4010 Wrist Supports

Catalog No. 19035275E Shop All Ergodyne Products
Change view
Click to view available options
Color:
Black
Tan
Size:
Small
Medium
Large
X-Large
Hand:
Left
Right
Model:
4000
4010
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size Hand Model
19-035275E Black Large Right 4010
19-035275D Black Medium Right 4010
19-035-271E Black Large Right 4000
19-035-271F Black X-Large Right 4000
19-035-271D Black Medium Right 4000
19-035272D Black Medium Left 4000
19-035272E Black Large Left 4000
19-035276C Black Small Left 4010
19-035276D Black Medium Left 4010
19-035-271C Black Small Right 4000
19-035273D Tan Medium Right 4000
19-035273E Tan Large Right 4000
19-035276E Black Large Left 4010
19-035276F Black X-Large Left 4010
19-035277C Tan Small Right 4010
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
This item is not returnable. View return policy
Catalog No. 19-035275E Supplier Ergodyne™ Supplier No. 70026
Add to Cart
Add to Cart
This item is not returnable. View return policy

Flexible support that doesn't compromise productivity

  • Ideal for material handling work
  • Durable supports encourage ergonomically neutral posture
  • Limit wrist flexion without compromising finger dexterity
  • Patented Open-Center Stay™ reduces pressure on median nerve
  • Fully adjustable for an exact fit and comfortable wear
  • Exclusive red line indicator ensures proper fit every time
  • Anti-odor treatment pulls moisture away from skin
  • Machine washable

ProFlex 4000

  • Limits downward wrist movement
  • ProFlex 4010
    • Adjustable double-closure stay that limits both upward and downward wrist movements

    Order Info

    Sold individually. Other styles available. See www.fishersafety.com.

    1 Measure around the smallest part of the wrist. This is a general guide only. Call your Fisher Safety Customer Service Representative at 1-800-772-6733 for more information.

    Specifications

    Color Black
    Closure Type Two stage hook and loop
    Product Type Proflex™ Single Strap Wrist Support
    Size Large
    For Use With (Application) Food processing, picking, packing, assembly, fabrication and taffy pulling
    Includes Patented open-center Stay™, red line indicator, anti-bacterial lining
    Hand Right
    Limitations Limits both upward and downward wrist movements
    Material Elastic
    Model 4010
    Size (English) 7 to 8 in.
    Size (Metric) 18 to 20 cm
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.