Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ ProFlex™ Thermal Utility Gloves

Catalog No. 19147340 Shop All Ergodyne Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-147-340 2X-Large
19-147-336 Small
19-147-337 Medium
19-147-338 Large
19-147-339 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-147-340 Supplier Ergodyne™ Supplier No. 16336
This item is not returnable. View return policy

Gloves are lined with 40 grams of 3M™ Thinsulate™ insulation for warmth, without compromising on dexterity.

  • Synthetic leather palm and fingers provide durability and comfort
  • Reinforced thumb and fingertips
  • Neoprene knuckle pad produces additional protection and wear
  • Woven elastic cuff with hook-and-loop closure
  • Machine washable

Specifications

Cuff Material Elastic
Coating Material Uncoated
Color Black
Hand Specific
Material Synthetic Leather
Product Type Mechanic's Gloves
Size 2X-Large
For Use With (Application) Cold Weather Work, Refrigeration, Construction, Mining, and Outdoor Recreation
Glove Cuff Style Hook and Loop
Liner Material 3M™ Thinsulate™
Series ProFlex™ 1633
Thumb Style Wing
Length (English) 11 in.
Length (Metric) 27.9 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.