Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ ProFlex™ 819WP Thermal Waterproof Gloves

Catalog No. 19147502 Shop All Ergodyne Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-147-502 Large
19-147-504 Small
19-147-503 Medium
19-147-501 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-147-502 Supplier Ergodyne™ Supplier No. 16044

Gloves feature a breathable waterproof and windproof Hipora liner, as well as 40 grams of 3M™ Thinsulate™ insulation. PVC coating on the palm, fingers, and saddle enhances grip capabilites in damp conditions.

  • Breathable waterproof and windproof Hipora insert liner
  • 3M™ Thinsulate™ thermal insulation for cold weather applications
  • PVC coating on the palm, fingers, and saddle enhances grip and durability
  • EVA knuckle pad with reflective dots for protection against bumps and bruises
  • Gauntlet cuff with pull on tab for easy donning and doffing, especially in wet conditions
  • Terrycloth brow wipe on the back of the thumb

Specifications

Back Material EVA Padding
Coated Area Palm, Fingers, Saddle
Coating Material PVC
Color Black
Hand Specific
Material Nylon, Hipora
Size Large
For Use With (Application) Cold Storage, Construction, Fishing, Ice Sculpting, Marine Research, and Winter Work
Glove Cuff Style Gauntlet
Liner Material 3M™ Thinsulate™ Insulation (40 g)
Series ProFlex™ 819WP
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.