Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ ProFlex™ 860 Lifting Gloves

Catalog No. 19050389 Shop All Ergodyne Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-050-389 X-Large
19-050-386 Medium
19-050-387 Large
19-050-395 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-050-389 Supplier Ergodyne™ Supplier No. 16125
This item is not returnable. View return policy

Gloves are made with a textured wet/dry gripping material for maximum surface area and optimal gripping effectiveness.

  • Half-finger design for dexterity
  • Durable, split leather padded palm
  • Breathable stretch spandex with neoprene knuckle pads
  • Woven elastic cuff with hook-and-loop closure
  • Terry thumb brow wipe

Specifications

Back Material Spandex
Cuff Material Elastic
Color Black
Hand Specific
Material Neoprene
Product Type Lifting Gloves
Size X-Large
Finger Style Half
For Use With (Application) Material Handling, Delivery, Packaging, Warehousing/Distribution, Moving, Shaking
Glove Cuff Style Hook and Loop
Series ProFlex™ 860
Length (English) 10 in.
Length (Metric) 25.4 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.