Learn More
Ergodyne™ ProFlex™ 9015F(x) Certified Anti-Vibration Gloves with Dorsal Protection
Shop All Ergodyne ProductsDescription
- Maximum shock, impact, and vibration protection with exclusive, patented Nu2O2™ polymer
- Pigskin leather palm and fingers for rugged wear and abrasion resistance
- PVC shell for extra protection for the back of the hand
- Finger tip reinforcement
- Micro-injected closure for quick cinching
- Woven elastic cuff for a perfect fit
- Wicked™ treated
- Pull-on tab
Specifications
Specifications
| Certifications/Compliance | ANSI S3.40, ISO 10819-1996 |
| Cuff Material | Woven Elastic |
| Color | Black |
| Hand | Specific |
| Material | Pigskin Leather |
| Product Type | Anti-vibration Gloves |
| Size | X-Large |
| For Use With (Application) | Demolition, Heavy Construction, Mining, Oil and Gas Refining |
| Glove Cuff Style | Open |
| Series | Proflex™ 9015F(x) |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.