Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ ProFlex™ 925F(x) Dorsal Impact-Reducing Gloves

Catalog No. 19147593 Shop All Ergodyne Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-147-593 X-Large
19-147-590 Small
19-147-591 Medium
19-147-592 Large
19-147-594 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-147-593 Supplier Ergodyne™ Supplier No. 16055
This item is not returnable. View return policy

Gloves offer a high level of protection with TPR armor on the knuckles, fingers, and carpal bone. Palm is reinforced with DuPont™ Aramid Fibers for extended wear and durability.

  • Molded TPR armor on knuckles, fingers and carpal bone provides impact protection, pinch injury prevention, and blow deflection
  • 5 mm EVA padding protects the metacarpal, palm, and ulnar regions
  • Abrasion-resistant PVC on palm and three fingers limits liquid penetration
  • Nonslip Armortex™ reinforcement zones on thumb saddle and index finger for enhanced grip and durability
  • Various flex zones in the armor for high dexterity
  • Reflective accents and hi-vis color scheme for optimal hand visibility
  • Contoured neoprene cuff with reflective pull-on tab resists snagging while offering a secure fit and easy on/off
  • Suitable for applications such as oil/gas drilling, extraction and refining, fracking, tool pushing, mining, demolition, heavy construction, and more

Specifications

Cuff Material Neoprene
Coating Material PVC
Color Hi-vis Lime Green, Hi-vis Orange, Black
Hand Specific
Material Thermoplastic Rubber
Product Type Anti-impact Gloves
Size X-Large
For Use With (Application) Oil/Gas Drilling, Extraction, Mining, Demolition, and Heavy Construction
Glove Cuff Style Open
Series ProFlex™ 925 F(x)
Length (English) 10 in.
Length (Metric) 25.4 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.