Learn More
Description
Ergoloid Mesylates USP is a white to off-white, microcrystalline or amorphous, practically odorless powder.
Alternative name(s): Ergoloid Mesilates, dihydroergotoxine mesilate,
Specifications
Specifications
| Chemical Name or Material | Ergoloid Mesylates USP |
| Packaging | Bottle, 15mL, Amber Glass Wide Mouth Packer, 28-400 |
| CAS | 8067-24-1 |
| Type | Drug Standard |
| Molecular Weight (g/mol) | 616.712 |
| Quantity | 1 g |
| Percent Purity | 97 to 103 % |
| Grade | USP |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
