Learn More
Impacto™ Ergomate Anti-fatigue Overshoes for Work boots
Impacto™ Ergomate Anti-fatigue Overshoes for Work boots are ideal for relief of tired, sore, feet, knees and back, improving safety and comfort. For areas where anti-fatigue matting is impractical due to length required or moving equipment.
Supplier: Impacto™ G88903S
Description
Impacto™ Anti-fatigue Ergomate Overshoes
- Layered EVA foam reduces impact and shock
- Allows the muscles to expand and contract naturally
- Improve circulation thus reducing pain and fatigue from walking and standing
- Patented adjustable strap system
- Suitable for all industries: warehouses, factories, assembly lines, machine shops, retail outlets, cashiers, clerks, laundry facilities, health care, postal workers, aircraft, warehouses and food processing
Specifications
CE Marked | |
EVA | |
Standard | |
Unisex | |
Ergomate Anti-fatigue Overshoes |
Black | |
Small; Men: 5 to 6, Women: 6 to 7 | |
Plain Toe | |
Yes |