Learn More
Labconco™ Ergonomic Chair
GSA_VASDP
Catalog No. 17200204 Shop All Labconco Corporation Products
Change view
Click to view available options
Product Type: Cleanroom Chair
Catalog No.
17-200-204 Product Type: Cleanroom Chair
1 options
Catalog No. 17-200-204 Supplier Labconco™ Supplier No. 3689700
Add to Cart
Add to Cart
Ergonomic Chair is a 4-way articulating seat for personalized comfort. ISO Class 6 clean room compatible.
  • 4-way articulating seat for personalized comfort
  • ISO Class 6 clean room compatible
  • Seat height adjusts from 19 to 26 in. (48 to 66 cm)
  • Five-leg composite base with resistance casters
  • 24.4 in. (62.0 cm) wide
  • Black vinyl upholstery
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Product Type Cleanroom Chair
With Casters Yes
With Tilt Yes
With Lumbar Support No
Color Black
With Backrest Yes
Autoclavable Not Autoclavable
Back Style Standard
With Armrest No
Footrest No
Material Vinyl
Seating Type Medium-form Chair