Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Surgical Design™ Ergonomical Surgical Knife Scalpel Handle

Catalog No. 22079796 Shop All Surgical Design Products
Change view
Click to view available options
Size:
# 3
# 4
# 8
Color:
Black
Blue
Green
Red
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Color
22-079-796 # 4 Red
22-079-797 # 8 Red
22-079-787 # 8 Green
22-079-816 # 8 Blue
22-079-783 # 4 Blue
22-079-791 # 3 Red
22-079-820 # 3 Green
22-079-824 # 3 Blue
22-079-826 # 3 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Catalog No. 22-079-796 Supplier Surgical Design™ Supplier No. 271PLRD
Add to Cart
Add to Cart
This item is not returnable. View return policy

Surgical Design™ Surgical Knife Scalpel Handle used for sharp and precise dissection.

  • Surgical Design™ Premium products exceed performance and quality expectations
  • Handles ergonomically designed for precision work
  • Comfortable, reusable
  • Blades slide on smoothly and securely
  • Superb quality and workmanship
  • Premium surgical grade stainless steel
  • Corrosion protected by passivation and ultrasonic cleaning
  • Sterilizable by EtOH/Gamma Radiation
  • Fully autoclavable

Specifications

Product Type Ergonomic Scalpel Handle
Handle Type Ergonomical
Size # 4
For Use With (Equipment) 20 Series Scalpel Blades
Disposable Reusable
Sterility Non-sterile
Certifications/Compliance ISO, GMP
Autoclavable Autoclavable
Handle Material PVC
Color Red
Graduated No
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.