Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Impacto™ Ultra Work Sport Anti-Fatigue Insoles

Impacto™ ERINWRK Ultra Work Sport Insoles help protect against back and knee pain and impact-related injuries such as plantar fasciitis, Achilles tendonitis, runner's knee and shin splints.

Supplier:  Impacto™ ERINWRKH

 View more versions of this product

Catalog No. 19-400-137


Only null left
Add to Cart

Description

Description

Impacto™ Ultra Work-Sport Anti-Fatigue Insoles

  • Sorbothane™ inlays absorb shock and help relieve pain in feet, knees and lower back
  • Designed to provide cushioned support and shock absorbing comfort
  • Cupped heel for rear foot stability
  • Breathable and antifungal
  • For use with closed shoes and boots
Specifications

Specifications

Blue
33.02 cm
Men: 14 to 15
Ergotech Ultra Work Sport Insoles
13 in.
Sorbothane
Unisex
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.