Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Foamtec ErgoWRENCH™ Vacuum Chamber Scrubbing Tools: Heads

Catalog No. 19304781 Shop All Foamtec International Products
Change view
Click to view available options
Product Type:
Fixed Scrub Head
Scrub Head
Size:
Large
Medium
Small
Square
Format:
Fixed
Non-removable, Fixed
Removable, Swivel
Swivel
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Size Format
19-304-781 Scrub Head Medium Removable, Swivel
19-304-780 Scrub Head Medium Non-removable, Fixed
19-304-779 Scrub Head Large Swivel
19-304-783 Fixed Scrub Head Small Non-removable, Fixed
19-304-778 Scrub Head Large Fixed
19-304-852 Scrub Head Square Fixed
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-304-781 Supplier Foamtec Supplier No. FT822R
This item is not returnable. View return policy

For safer and more efficient scrubbing of hard to reach areas throughout vacuum chambers

Stiff swivel head can be adjusted to optimize scrubbing angles. Different size heads are interchangeable, which allows the ErgoWRENCH to be adjusted for different applications.

  • Reduces scrub-time by focusing Diamond ScrubPAD directly onto heavy deposition buildup, quickly taking chamber back to bare metal
  • Removes heavy deposition from areas critical to production, preventing unwanted flaking
  • Safely removes deposition from sensitive areas
  • Quick removal of deposition from aluminum, stainless-steel, ceramic, glass, quartz and anodized surfaces while minimizing tool wear

Specifications

Product Type Scrub Head
Format Removable, Swivel
For Use With (Application) For safer and more efficient scrubbing of hard to reach areas
For Use With (Equipment) Interchangeable handles
Includes Hook
Material Delrin™ Plastic
Size Medium
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.