Learn More
Description
Stiff swivel head can be adjusted to optimize scrubbing angles. Different size heads are interchangeable, which allows the ErgoWRENCH to be adjusted for different applications.
- Reduces scrub-time by focusing Diamond ScrubPAD directly onto heavy deposition buildup, quickly taking chamber back to bare metal
- Removes heavy deposition from areas critical to production, preventing unwanted flaking
- Safely removes deposition from sensitive areas
- Quick removal of deposition from aluminum, stainless-steel, ceramic, glass, quartz and anodized surfaces while minimizing tool wear
Specifications
Specifications
| Product Type | Fixed Scrub Head |
| Format | Non-removable, Fixed |
| For Use With (Application) | For safer and more efficient scrubbing of hard to reach areas |
| For Use With (Equipment) | Interchangeable handles |
| Includes | Hook |
| Material | Delrin™ Plastic |
| Size | Small |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.