Learn More
Medchemexpress LLC HY-13442A 5mg , Eribulin (mesylate) CAS:441045-17-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY13442A5MG
Encompass_Preferred
HY-13442A 5mg Eribulin (mesylate) CAS: 441045-17-6 Eribulin mesylate (E7389 mesylate) is a microtubule targeting agent that is used for the research of metastatic breast cancer. Eribulin mesylate inhibits the proliferation of cancer cells by binding microtubule proteins and microtubules. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9702
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.