Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eriochrome Black T, ACS Reagent (EBT; Mordant Black 11), Reagents

Catalog No. 01336764
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-336-764 25 g
01-336-766 100 g
01-336-765 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-336-764 Supplier Reagents Holdings Llc Supplier No. C611020025C1
Add to Cart
Add to Cart

Eriochrome Black T, ACS Reagent (EBT; Mordant Black 11), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 1787-61-7
Molecular Formula C20H12N3NaO7S
Molecular Weight (g/mol) 461.38
InChI Key JHUJLRKQZAPSDP-GXTSIBQPSA-M
Synonym Water hardness indicator
IUPAC Name sodium (4Z)-4-[2-(1-hydroxynaphthalen-2-yl)hydrazin-1-ylidene]-7-nitro-3-oxo-3,4-dihydronaphthalene-1-sulfonate
SMILES [Na+].OC1=C2C=CC=CC2=CC=C1N\N=C1/C(=O)C=C(C2=CC(=CC=C12)[N+]([O-])=O)S([O-])(=O)=O

Specifications

Boiling Point 1413°C
Grade ACS
Packaging Glass Bottle
Specific Gravity 1.109
Color Black to Brown
Quantity 25 g
Physical Form Solid
Chemical Name or Material Eriochrome Black T, ACS Reagent (EBT, Mordant Black 11)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.