Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eriochrome™ Black T, in Methoxyethanol, for Hardness, Certified, LabChem™

Catalog No. LC140401
Change view
Click to view available options
Quantity:
125 mL
500 mL
1 L
Packaging:
Amber Glass
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC140401 500 mL Amber Glass
LC140407 125 mL Amber Glass
LC140402 1 L Amber Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. LC140401 Supplier LabChem, Inc. Supplier No. LC140401
Add to Cart
Add to Cart

Chemical Identifiers

CAS 109-86-4 , 1787-61-7
Molecular Formula C3H8O2
Molecular Weight (g/mol) 76.095
InChI Key XNWFRZJHXBZDAG-UHFFFAOYSA-N
PubChem CID 8019
ChEBI CHEBI:46790
IUPAC Name 2-methoxyethanol
SMILES COCCO

Specifications

CAS Max % 99.5, 0.5
Linear Formula C20H12N3O7NaS
UN Number UN1188
Solubility Information Soluble in water
Identification Passes Test
Formula Weight 461.38
Grade Certified
Packaging Amber Glass
Suitability Passes Test
Decomposition Information Carbon monoxide; Carbon dioxide; May release flammable gases
Color Maroon
Quantity 500 mL
Physical Form Liquid
Chemical Name or Material Eriochrome™ Black T, in Methoxyethanol, for Hardness
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Flammable liquid and vapor.
Harmful if swallowed, in contact with skin or if inhaled.
May damage fertility or the unborn child.
Precautionary Statement GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Keep away from heat, sparks, open flames, hot surfaces.
No smoking.
Keep container tightly closed.
Ground/bond container and receiving equipment.
Use explosion-proof electrical, ventilating, lighting equipment.
Use only non-sparking tools.
Take precautionary measures against static discharge.
Avoid breathing mist.
Do not eat, drink or smoke when using this product.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
Use only outdoors or in a well-ventilated area.
If exposed or concerned: Get medical advice/attention.
If swallowed: Rinse mouth.
Call a poison center/doctor if you feel unwell.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Call a poison center/doctor if you feel unwell.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
Store locked up in a cool, well-ventilated place.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.