Learn More
Eriochrome™ Blue-Black R, Dry Form in NaCl, for Calcium, Certified, LabChem™
Shop All LabChem ProductsSpecifications
Specifications
| Melting Point | 801°C |
| Boiling Point | 1413°C |
| CAS Max % | 99.8, 0.2 |
| Identification | Passes Test |
| Packaging | Poly Bottle |
| Specific Gravity | 2.165g/cm3 |
| Suitability | Passes Test |
| Linear Formula | HOC10H6N:NC10H5OHSO3H |
| Decomposition Information | Hydrogen chloride; Carbon dioxide; Carbon monoxide; Nitrogen oxides; Sulfur compounds |
| Density | 2.165g/cm3 |
| Show More |
Safety and Handling
| Hazard Statement | GHS H Statement Product is not hazardous. |
| Precautionary Statement | GHS P Statement If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes. If swallowed, get medical attention. |
| EINECSNumber | 231-598-3 |
| Recommended Storage | Room Temperature |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.