Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eriochrome Blue Black R Indicator, 0.2% (w/w) in Sodium Chloride, Ricca Chemical
SDP

Catalog No. R2902000200 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
100 g
200 g
500 g
Packaging:
Natural Poly Bottle
White Poly Wide Mouth Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R2902000200 200 g White Poly Wide Mouth Bottle
2902-100 100 g White Poly Wide Mouth Bottle
R2902000500 500 g Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. R2902000200 Supplier Ricca Chemical Company Supplier No. 2902200
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2538-85-4 , 7647-14-5
Molecular Formula C20H13N2NaO5S
Molecular Weight (g/mol) Mixture
InChI Key YJUGDMOGPJJWDG-UHFFFAOYSA-M
PubChem CID 23702480
IUPAC Name sodium;4-[(2-hydroxynaphthalen-1-yl)hydrazinylidene]-3-oxonaphthalene-1-sulfonate
SMILES C1=CC=C2C(=C1)C=CC(=C2NN=C3C4=CC=CC=C4C(=CC3=O)S(=O)(=O)[O-])O.[Na+]

Specifications

Concentration 0.2% (w/w)
Packaging White Poly Wide Mouth Bottle
Solvent or Matrix Sodium Chloride
Color Purple/Violet
Quantity 200 g
Physical Form Powder
Grade Indicator
Chemical Name or Material Eriochrome Blue Black R Indicator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.