Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Multispecies AKT/ERK/p70 S6K Activation InstantOne™ ELISA Kit

Catalog No. 50246235
Encompass_Preferred
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-246-235 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-246-235 Supplier Invitrogen™ Supplier No. 858601811
Add to Cart
Add to Cart

ELISA

The InstantOne™ ELISA is specifically engineered for accurate measurement of phosphorylated ERK 1/2 (human, mouse, hamster), AKT 1/2/3 (human, mouse, hamster), and p70 S6K (human, mouse) in cell lysates. Detection of these phosphorylated molecules in rat is also expected. The InstantOne™ ELISA kit allows for fast analysis of samples in approximately one hour. All reagents used in a traditional sandwich ELISA are added in solution to a plate followed by a wash step and detection with the TMB colorimetric substrate.

Extracellular signal-regulated kinase (ERK) is the founding member and a key component of the classical Mitogen-Activated Protein Kinase (MAPK) pathway. MEK activates ERK 1/2 by phosphorylation of its TxY motifs in response to a variety of stimuli, including growth factors and cytokines. AKT, which exists as multiple isoforms, is one of the principle kinases activated by phosphoinositide 3-kinase (PI3K). These lipid second messengers bind to the pleckstrin homology domain of AKT to promote its translocation to the plasma membrane for activation via phosphorylation at Thr308 and Ser473 by PDK1 and the mTOR TORC2 complex, respectively. Phosphorylation at both these sites is required for full activation of AKT Ser/Thr kinase activity. p70 S6 Kinase (p70S6K) is a member of the ribosomal S6 kinase family of Ser/Thr kinases. p70 S6K activity is controlled by multiple signaling pathways, including the MAPK, phosphoinositide-3 kinase (PI3K), and mTOR pathways. This regulation results in the phosphorylation of multiple sites of p70 S6K, including Thr389 within the linker domain, which is required for full activation. Activated p70 S6K phosphorylates several residues on the S6 ribosomal protein, thereby leading to increased protein synthesis.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P23443,P27361,P28482,P31749,P31751,Q9Y243
Conjugate HRP
Product Type ELISA
Sample Type Cell Lysates
Target Species Human
For Use With (Application) ELISA
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 10000, 207, 208, 5594, 5595, 6198
Gene Symbol AKT,CWS6,PKB,PKB-ALPHA,PRKBA,RAC,RAC-ALPHA,ERK,ERK-2,ERK2,ERT1,MAPK2,P42MAPK,PRKM1,PRKM2,p38,p40,p41,p41mapk,p42-MAPK,PS6K,S6K,S6K-beta-1,S6K1,STK14A,p70 S6KA,p70(S6K)-alpha,p70-S6K,p70-alpha,ERK-1,ERK1,ERT2,HS44KDAP,HUMKER1A
Kit Contents Capture Antibody, Detection Antibody Reagent, InstantOne™ Assay Plate, Cell Lysis Buffer, Enhancer Solution, Wash Buffer (10X), Substrate Reagent (TMB), Stop Solution, Positive Control Cell Lysate (lyophilized), Plate seal
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Cell Lysate 20 μL
Storage Requirements 2°C to 8°C
Target AKT Pan/Erk1/2/p70 S6 Kinase
Test Time 1 hrs 20 mins
Total Assay Time 1 hrs 10 mins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.