Learn More
ERK3 Mouse anti-Human, Clone: OTI5E1, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF500077

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
ERK3 is a member of the Ser/Thr protein kinase family, and is most closely related to mitogen-activated protein kinases (MAP kinases). MAP kinases also known as extracellular signal-regulated kinases (ERKs), are activated through protein phosphorylation cascades and act as integration points for multiple biochemical signals. This kinase is localized in the nucleus, and has been reported to be activated in fibroblasts upon treatment with serum or phorbol esters.Specifications
ERK3 | |
Monoclonal | |
Unconjugated | |
MAPK6 | |
ERK3, HsT17250, PRKM6, p97MAPK | |
Mouse | |
Affinity Chromatography | |
RUO | |
5597 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI5E1 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q16659 | |
MAPK6 | |
Recombit protein expressed in E.coli corresponding to amino acids 345-721 of human ERK3 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.