Learn More
United Scientific™ Erlenmeyer Flask, 24/40 Joint
SDPSCI_ED
Catalog No. S16095 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Capacity (Metric):
125 mL
250 mL
300 mL
500 mL
1000 mL
2000 mL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
S16095 300 mL
S16094 125 mL
S15998 500 mL
S15733 250 mL
S15737 2000 mL
S16048 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. S16095 Supplier United Scientific™ Supplier No. UNJTERFL300
Add to Cart
Add to Cart
Made from ASTM E-438 Type-1 Class A, Borosilicate 3.3 glass
  • Borosilicate glass flasks are designed to give the best combination of thermal shock resistance and physical strength.
  • Feature 24/40 jointed sockets and graduations in durable white enamel.
  • Made from ASTM E-438 Type-1 Class A, Borosilicate 3.3 glass.
  • These flasks comes with ground joints manufactured as per ASTM E-1404, Type 2, Class-1 requirement.
  • The flasks are printed in white color enamel.
Specifications
Capacity (Metric) 300 mL
Material Borosilicate Glass (3.3)
Autoclavable Autoclavable
Disposable Reusable
Graduated Yes
Mouth Style Joint
Sterility Non-sterile
Bottom Shape Flat
Closure Type 24/40 Joint
Surface Treatment No