Learn More
ERMN Mouse anti-Human, Mouse, Porcine, Rat, Clone: 2B1C1, Proteintech
Shop All Proteintech Group Inc Products

Description
The predicted MW of ERMN is 33 kDa, while western blot analyses of different tissue lysates using 66605-1-Ig antibody to ERMN detected a 50 kDa single band protein as a result of phosphorylation modification.
Specifications
Specifications
| Antigen | ERMN |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | 2B1C1 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 |
| Gene | ERMN |
| Gene Accession No. | Q5EBJ4, Q5RJL0, Q8TAM6 |
| Gene Alias | ermin, ermin, ERM like protein, ERMN, JN, Juxtanodin, KIAA1189 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.