Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ERMP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5140082
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5140082 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5140082 Supplier Invitrogen™ Supplier No. PA5140082
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: LGLALYLIAL RTLVQLSLQQ LVLRGAAGHR GEFDALQARD YLEHITSIGP RTTGSPENEI LTVHYLLEQI KLIEVQSNSL HKISVDVQRP TGSFSIDFLG GFTSYYDNIT NVVVKLEPRD GAQHAVLAN Highest antigen sequence indentity to the following orthologs: Rat - 84%, Mouse - 84%.

Within the ovary, required for the organization of somatic cells and oocytes into discrete follicular structures.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ERMP1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ERMP1
Gene Accession No. Q7Z2K6
Gene Alias aminopeptidase Fxna; bA207C16.3; bA207C16.3 (novel protein similar to predicted yeast, plant and worm proteins); D19Ertd410e; D19Wsu12e; Endoplasmic reticulum metallopeptidase 1; Ermp1; Felix-ina; Fxna; KIAA1815; mKIAA1815; putative aminopeptidase Fxna
Gene Symbols ERMP1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ERMP1. Recombinant protein control fragment (Product #RP-108921).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79956
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.