Learn More
TARGETMOL CHEMICALS INC BOCEPREVIR 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T4988
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Boceprevir (SCH 503034) is a novel potent highly selective orally bioavailable HCV NS3 protease inhibitor with Ki of 14 nM in both enzyme assay and EC90 of 350 nM in cell-based replicon assay. purity: 99%
Catalog No. 50-257-9843
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.