Learn More
Selleck Chemical LLC 4-Bromo-3-hydroxybenzoic acid S5444-25mg
SDP
Supplier:  Selleck Chemical LLC S544425MG
Encompass_Preferred
4-bromo-3-hydroxybenzoic acid is a potent inhibitor of the enzyme histidine decarboxylase with resultant inhibition of histamine formation in mammals
Catalog No. 50-310-9516
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.