Learn More
Description
- Determine ABO/Rh blood type of real human blood in under five minutes.
- A small amount of blood is applied to a series of wells containing lyophilized blood typing antibodies, followed by the addition of a wash buffer that flows across each well.
- Red blood cells that agglutinate in the presence of a particular antibody will not be rinsed from the sample well and the well will remain red, indicating a positive result.
- Red blood cells that have not undergone agglutination will be rinsed from the well and the well will revert to white, indicating a negative result.
- Each card contains a negative control well to ensure accurate results. Kit contains enough materials for 1 test. Instructions are included.
- Kit Includes:
- 6 mL Buffer Solution
- 1 Erycard™
- 1 Alcohol Wipe
- 1 Lancet
- 4 Blood Collection Sticks
Specifications
Specifications
| CE Marker | No |
| Detectable Analytes | ABO Antigen, Rh D antigen |
| Type | Card |
| For Use With (Application) | Educational/Instructional Laboratory Use Only |
| Includes | 6 mL Buffer Solution, 1 Erycard, 1 Alcohol Wipe, 1 Lancet, 4 Blood Collection Sticks |
| Test Time | 5 Minutes |
| Form | Ready-to-Use (RTU) |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
