Learn More
Bioworld Erythrina cristagalli (Coral tree) Lectin (ECL/ECA) - Texas Red
Supplier:  Bioworld 217610202
Encompass_Preferred
Technical Specifications:Ligand Source:Erythrina cristagalliMolecular Weight:54 kDaCarbohydrate Specificity:Galβ1,4GlcNAcInhibiting/Eluting Sugar:LactoseCarbohydrate for Elution:0.2MLactoseBlood Group Specificity:A (-SA)Conjugate: texas RedForm:LiquidIsoelectric (pI):6.3-6.5Storage: -20°CApplication note: Calcium, Manganese, and Zinc are required for binding.
Catalog No. 50-199-126
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.