Learn More
United States Biological Corporation Erythromycin A 9,11-Imino Ether, 161193-44-8
Catalog No. NC1310544 Shop All United States Biological Corporation Products
Encompass_Preferred
Molecular Formula C37H66N2O12, Purity ≥95%, Molecular Weight 730.93, Melting Point >134ºC (dec.), Solubility: Chloroform (slightly), DMSO (Slightly) Methanol (slightly). Erythromycin A 9,11-Imino Ether is an impurity in the synthesis of Erythromycin, a macrolide antibiotic with broad spectrum of antibacterial activity.
Catalog No. NC1310544 Supplier United States Biological Corporation Supplier No. 1640412.5MG
Molecular Formula C37H66N2O12, Purity ≥95%, Molecular Weight 730.93, Melting Point >134ºC (dec.), Solubility: Chloroform (slightly), DMSO (Slightly) Methanol (slightly). Erythromycin A 9,11-Imino Ether is an impurity in the synthesis of Erythromycin, a macrolide antibiotic with broad spectrum of antibacterial activity.