Learn More
Abcam Erythromycin, Macrolide Antibiotic, Solid, 733.9 Da, 1 g
Description
- Molecular Weight: 733.9 Da
- Form: Solid
- Mechanism of Action: Macrolide antibiotic that inhibits cytochrome p450 3A4 and P-glycoprotein. Binds the 50S ribosomal subunit to inhibit protein synthesis in Gram-positive and Gram-negative bacteria.
- Composition: 100% Erythromycin
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Reactivity | Gram-positive and Gram-negative bacteria |
| Shipping Condition | Ambient |
| Target Molecule | 50S Ribosomal Subunit, Cytochrome P450 3A4, P-glycoprotein |
| Form | Solid |
| Buffer | 100% Erythromycin |
| Content And Storage | Store at 4°C. |
| Quantity | 1 g |
| For Use With (Application) | Antibiotic research, Protein synthesis inhibition, Cytochrome p450 3A4 inhibition, P-glycoprotein inhibition |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.