Learn More
FUJIFILM Wako PYROSTAR™ ES-F 80 Test Kit
Description
- Endotoxin-specific reagent avoids false positive results from glucans
- Dual-purpose reagent can perform either gel clot or kinetic turbidimetric assay (KTA)
- Gel clot sensitivities range from 0.015 to 0.25 EU/mL
- Kinetic turbidimetric sensitivities range from 0.001 to 10 EU/mL
- Chances of interference are reduced due to high sensitivity
- Facilitates easier transition from gel clot to KTA
Specifications
Specifications
| Form | Lyophilized |
| Fill Volume | 2 mL |
| Includes | 4 x 2 mL multi-test vials and 1 vial CSE (500 ng/vial) |
| Quantity | 80 Tests |
| Sensitivity | 0.06 EU/mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.