Learn More
ATCC Escherichia coli (Migula) Castellani and Chalmers, ATCC BAA-3310, strain 1013491
Supplier: ATCC BAA3310
Escherichia coli (Migula) Castellani and Chalmers, strain 1013491, is a clinically derived, drug-resistant isolate recovered from a human urine sample in Greece in 2013. It is supplied as a frozen research culture intended for microbiology and antimicrobial resistance studies.
- Clinically derived isolate from a human urine sample.
- Strain designation 1013491.
- Isolated in Greece in 2013 from a 54-year-old female.
- Drug-resistant isolate suitable for antimicrobial resistance research.
- Supplied as a frozen culture; store at -80 °C or colder.
- Recommended growth on Trypticase Soy Agar or broth.
- Incubate aerobically at 37 °C.