Learn More
TRIBIOSCIENCE INC Escherichia coli (E. Coli) - Salmonella qPCR Kit, 200 Tests
SDP
Supplier:  TRIBIOSCIENCE INC TBS42051200
Encompass_Preferred
The Escherichia Coli (E. Coli)-Salmonella Multiple qPCR Kit provides a fast, accurate, and simple approach to detecting E. coli and salmonella contamination of food ingredients, cannabis products, water, environmental samples, or other biosamples.
Catalog No. NC2887502