Learn More
ATCC Escherichia coli K-12 RV308 Freeze-dried Culture (ATCC 31608)
Supplier:  ATCC 31608
Encompass
Escherichia coli K-12 RV308 is a laboratory reference strain supplied in a freeze-dried format for reconstitution and propagation. It is intended for research, strain characterization, and molecular biology workflows and is provided with recommended storage guidance and usage notes.
  • Freeze-dried for long-term stability.
  • Genome sequenced strain.
  • Supplied for reconstitution and propagation in laboratory media.
  • Recommended storage conditions: 2°C to 8°C.
  • Suitable for strain characterization, molecular biology, and comparative genomics.
  • Patent-deposit material; follow applicable use restrictions.
Catalog No. NC3844737