Learn More
ATCC Escherichia coli (Migula) Castellani and Chalmers - ATCC BAA-2201 Freeze-dried Research Strain, Strain 2002-3092, BSL-2
Supplier:  ATCC BAA2201
Encompass
Escherichia coli (Migula) Castellani and Chalmers (strain 2002-3092) is a freeze-dried, toxigenic research strain confirmed by PCR to carry eae and stx1 (stx1+, stx2-, eae+). Isolated from stool, the culture is supplied with rehydration and culture instructions and is intended for laboratory research applications including enteric disease studies, food testing, and infectious disease research. It must be handled under BSL-2 conditions and is not for therapeutic, diagnostic, or consumption use.
  • Freeze-dried format for convenient storage and transport.
  • PCR-confirmed genotype: stx1+, stx2-, eae+.
  • Suitable for enteric disease, food testing, and infectious disease research.
  • Storage at 2°C to 8°C; consider vapor-phase liquid nitrogen for long-term preservation.
  • Culture conditions: nutrient agar or nutrient broth; incubate at 37°C, aerobic.
  • BSL-2 handling required; use appropriate personal protective equipment.
Catalog No. NC2964375