Learn More
ATCC Escherichia coli (Migula) Castellani and Chalmers - ATCC BAA-3303, drug-resistant bacterial strain (frozen vial)
Supplier:  ATCC BAA3303
Encompass
Escherichia coli clinically derived strain isolated from a human pancreas in 2014. The strain is genome sequenced and characterized for multiple antimicrobial resistance determinants; it is supplied frozen and must be handled under BSL-2 laboratory conditions. Common uses include antimicrobial resistance research, molecular assay development, and antimicrobial drug discovery.
  • Clinically derived, isolated from human pancreas in 2014.
  • Genome sequenced and annotated for multiple resistance genes.
  • Phenotypically resistant to multiple commonly used antibiotics; includes a documented susceptibility profile.
  • Supplied as a frozen vial; recommended storage at -80°C or colder.
  • Recovery: incubate at 37°C for 18-24 hours on appropriate media.
  • Handle under biosafety level 2 (BSL-2) containment and follow institutional safety protocols.
Catalog No. NC3538765