Learn More
ATCC Escherichia coli (Migula) Castellani and Chalmers Strain NIH RZ1032 (ATCC 39737) - Frozen Culture, Transformation Host
Supplier:  ATCC 39737
Encompass
Escherichia coli (Migula) Castellani and Chalmers, strain NIH RZ1032, is supplied as a frozen bacterial culture for laboratory use. This strain is used as a transformation host and supports propagation of filamentous phage M13mp8; it can produce M13mp2 phage that contains uracil in the DNA. The product is intended for BSL-1 laboratory applications and microbiology or molecular biology workflows.
  • Organism: Escherichia coli (Migula) Castellani and Chalmers
  • Strain designation: NIH RZ1032
  • Product format: Frozen culture
  • Recommended medium: LB agar/broth (Miller)
  • Incubation temperature: 37°C
  • Biosafety level: BSL 1
  • Typical applications: Transformation host; phage propagation
  • Notes: Material deposited with a patent depository; mission collection item
Catalog No. NC3453730