Learn More
Medchemexpress LLC HY-N0632 5mg Medchemexpress, Esculentoside A CAS:65497-07-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN06325MG
Encompass_Preferred
Medchemexpress, HY-N0632 5mg Esculentoside A CAS:65497-07-6 Esculentoside A (EsA), a kind of triterpene saponin isolated from roots of Phytolacca esculenta.Esculentoside A (EsA) possesses anti-inflammatory activity in acute and chronic experimental models, has selective inhibitory activity towards cyclooxygenase-2 (COX-2).Esculentoside A (EsA) suppresses inflammatory responses in LPS-induced acute lung injury (ALI) through inhibition of the nuclear factor kappa B (NF-ΚB) and mitogen activated protein kinase (MAPK) signaling pathways. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2806
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.