Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Esculin, Spectrum™ Chemical
SDP

Catalog No. 18620749 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
5 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-620-749 100 g
18-620-747 5 g
18-620-748 25 g
18-620-750 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-620-749 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. ES108100GM
Add to Cart
Add to Cart

Synonym: Aesculin, Esculoside, Polychrome, 6,7-Dihydroxycoumarin-6-Glucoside

Esculin is also known as aesculin and occurs naturally in the horse chestnut. It is used in a microbiology laboratory to aid in the identification of bacterial species Ungraded products supplied by Spectrum™ are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Chemical Identifiers

CAS 531-75-9

Specifications

Quantity 100 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.