Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ ESI 05
GREENER_CHOICE

Catalog No. p-7146000
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
63-205 5 mg
63-202-5 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 63-205 Supplier Tocris Bioscience™ Supplier No. 6320/5
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Epac2 inhibitor

ESI 05 is an EPAC2 inhibitor. Inhibits cAMP binding to EPAC2 and cAMP mediated EPAC2 guanine nucleotide exchange factor (GEF) activity (IC50 = 0.4 μM).

Specifications

Chemical Name or Material 1,3,5-Trimethyl-2-[(4-methylphenyl)sulfonyl]benzene
CAS 5184-64-5
Quantity 5 mg
Molecular Formula C16H18O2S
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.