Learn More
Amsbio LLC Esophagus squamous cell carcinoma with matched cancer adjacent tissue and esophagus lymph node tissue array/ including pathology grade/ TNM and clinical stage/ 50 cases/150 cores(core size 1.5mm)/ replacing ES1505
Supplier:  Amsbio LLC ES1505A
Encompass
Esophagus squamous cell carcinoma with matched cancer adjacent tissue and esophagus lymph node tissue microarray/ containing 17 cases of squamous cell carcinoma and matched cancer adjacent tissue and esophagus lymph node tissue/ 33 squamous cell carcinoma with matched cancer adjacent tissue and lymph node metastatic squamous cell carcinoma/ triplicate cores per case
Catalog No. NC3007567