Learn More
US Biomax Inc Esophagus squamous cell carcinoma with matched adjacent normal esophagus tissue array, including pathology grade, TNM and clinical stage, 60 cases/120 cores, replacing ES1201
SDP
Supplier:  US Biomax Inc ES1202
Encompass
Esophagus squamous cell carcinoma with matched adjacent normal esophagus tissue microarray, containing 59 cases of squamous cell carcinoma and and matched adjacent esophagus tissue, 1 carcinosarcoma and matched adjacent esophagus tissue, duplicate cores per case
Catalog No. NC2958570
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.