Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ESRRA Monoclonal Antibody (OTI2G5), TrueMAB™, OriGene
SDP

Catalog No. 501682574
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-2574 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-2574 Supplier OriGene Supplier No. TA809112
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Estrogen-related receptor alpha (ERR alpha), a NR3 Steroid Receptor, was isolated based on sequence similarity in its DNA-binding domain to estrogen receptor alpha (ER alpha). ERR alpha has been shown to regulate the promoters of lactoferrin, medium-chain acyl CoA dehydrogenase, osteopontin, and thyroid receptor alpha, and it may affect cellular energy balance and bone formation.

Specifications

Antigen ESRRA
Applications Western Blot
Classification Monoclonal
Clone OTI2G5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene ESRRA
Gene Accession No. P11474
Gene Alias ERR1, ERRa, ERRalpha, ESRL1, NR3B1
Gene Symbols ESRRA
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 206-417 of human ESRRA produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2101
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.