Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ESRRA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA1314 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA1314 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA1314 Supplier Invitrogen™ Supplier No. PA1314
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PA1-314 detects human ERR1 from transfected cells and shows no cross-reactivity to ERR3. PA1-314 has been successfully used in Western blot and immunohistochemistry procedures. By Western blot, this antibody detects an ~52 kDa protein representing ERR1 from COS-7 transfected cells. An unidentified band is seen at ~37 kDa. The PA1-314 immunogen is a synthetic peptide corresponding to the residues M(1) S S Q V V G I E P L Y I K A E(16) of human ERR1. This sequence is completely conserved between canine, human, mouse and rat. This immunizing peptide (PEP-231) can be used for neutralization and control experiments.

Estrogen-related receptor alpha 1 (ERR alpha, ERR1) is an orphan member of the nuclear hormone receptor superfamily. All the ERR family members share an almost identical DNA-binding domain, which has 68% amino acid identity with that of estrogen receptor. This protein binds to an ERR-alpha response element (ERRE) containing a single consensus half-site, 5'-tnaaggtca-3' can bind to the medium-chain acyl coenzyme a dehydrogenase (mcad) response element nrre-1 and may act as an important regulator of mcad promoter. Studies have shown that ERR1 functions a metabolic regulator and is often expressed in tissues that preferentially metabolize fatty acids. The corresponding gene for the ERR1 is NR3B1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ESRRA
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA and 0.05% sodium azide
Gene ESRRA
Gene Accession No. P11474, Q5QJV7
Gene Alias ERR alpha; ERR1; ERRa; ERRalpha; ERR-alpha; Errra; ESRL1; ESRRA; ESRRA1; ESRRA2; estrogen receptor related 1; estrogen receptor-like 1; estrogen related receptor alpha; estrogen related receptor alpha 1; estrogen related receptor, alpha; estrogen-related nuclear receptor alpha; estrogen-related receptor alpha; estrogen-related receptor, alpha; Estrra; HERR1; LOW QUALITY PROTEIN: steroid hormone receptor ERR1; NR3B1; Nuclear receptor subfamily 3 group B member 1; orphan nuclear receptor; steroid hormone receptor ERR1; Steroid hormone receptor ERR1 (Estrogen-related receptor, alpha) (ERR-alpha) (Estrogen receptor-like 1)
Gene Symbols ESRRA
Host Species Rabbit
Immunogen Synthetic peptide corresponding to the residues M(1) S S Q V V G I E P L Y I K A E(16) of human ERR1.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2101, 293701
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.